Human ATP-binding Cassette Sub-family A Member 3 ELISA Kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) design ed for the in vitro quantitative determination of human ATP-binding Cassette Sub-family A Member 3 in serum, plasma, 1 * 96 Tests
Catalog # ANTIA310735-96
Supplier: ANTIBODIES.COM