ContactUsLinkComponent
This Superoxide anion Scavenging Capacity Assay Kit is a quantitative assay kit for measuring Superoxide Anion Scavenging Capacity i n animal and plant tissues, cells, and blood. 1 * 48 Tests
Catalog # ANTIA319740-48
Supplier:  Antibodies.com
New Product
This Superoxide anion Scavenging Capacity Assay Kit is a quantitative assay kit for measuring Superoxide Anion Scavenging Capacity i n animal and plant tissues, cells, and blood. 1 * 48 Tests
Catalog # ANTIA319740-48
Supplier:  Antibodies.com