Rabbit TNF alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rabbit TNF alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologi 1 * 48 Tests
Catalog # ANTIA2996-48
Supplier: Antibodies.com