ContactUsLinkComponent
This gamma Glutamyl Cysteine Ligase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Cysteine Ligase acti vity in serum, plasma, animal and plant tissues, cells, bacteria, and fungi. 1 * 48 Tests
Catalog # ANTIA319760-48
Supplier:  ANTIBODIES.COM
New Product
This gamma Glutamyl Cysteine Ligase Activity Assay Kit is a quantitative assay kit for measuring gamma Glutamyl Cysteine Ligase acti vity in serum, plasma, animal and plant tissues, cells, bacteria, and fungi. 1 * 48 Tests
Catalog # ANTIA319760-48
Supplier:  ANTIBODIES.COM