Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human 5HT1A Receptor protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA325073-10
Supplier: ANTIBODIES.COM