ContactUsLinkComponent
This Tissue and Blood Alkaline Phosphatase Activity Assay Kit is a quantitative assay kit for measuring Alkaline Phosphatase activit y in serum, plasma, animal tissues, and urine. 1 * 48 Tests
Catalog # ANTIA319746-48
Supplier:  ANTIBODIES.COM
New Product
This Tissue and Blood Alkaline Phosphatase Activity Assay Kit is a quantitative assay kit for measuring Alkaline Phosphatase activit y in serum, plasma, animal tissues, and urine. 1 * 48 Tests
Catalog # ANTIA319746-48
Supplier:  ANTIBODIES.COM