ContactUsLinkComponent
Human Myelin Oligodendrocyte Glycoprotein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Myelin Oligodendrocyte Glycoprotein in tissue homogenates, cell lysates or oth 1 * 48 Tests
Catalog # ANTIA3149-48
Supplier:  ANTIBODIES.COM
New Product
Human Myelin Oligodendrocyte Glycoprotein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Myelin Oligodendrocyte Glycoprotein in tissue homogenates, cell lysates or oth 1 * 48 Tests
Catalog # ANTIA3149-48
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.