ContactUsLinkComponent
This one-step Dewaxing/Antigen Retrieval Buffer is a rapid reagent for paraffin slides pretreatment. There is no need to do the trad itional three times of xylene dewaxing, three to five times of gradient ethanol hydration, all shorten to one solutio 1 * 200 mL
Catalog # ANTIA293491-200
Supplier:  ANTIBODIES.COM
New Product
This one-step Dewaxing/Antigen Retrieval Buffer is a rapid reagent for paraffin slides pretreatment. There is no need to do the trad itional three times of xylene dewaxing, three to five times of gradient ethanol hydration, all shorten to one solutio 1 * 200 mL
Catalog # ANTIA293491-200
Supplier:  ANTIBODIES.COM