ContactUsLinkComponent
Human Anti-IL-18BP Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Anti-IL-18BP Antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, and 1 * 96 Tests
Catalog # ANTIA326373-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-IL-18BP Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Anti-IL-18BP Antibody in serum, plasma, cell culture supernatant, cell or tissue lysate, and 1 * 96 Tests
Catalog # ANTIA326373-96
Supplier:  ANTIBODIES.COM