ContactUsLinkComponent
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Beta B.1.351 Variant) containing the following mutations : K417N, E484K, N501Y. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318199-100
Supplier:  ANTIBODIES.COM
New Product
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Beta B.1.351 Variant) containing the following mutations : K417N, E484K, N501Y. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA318199-100
Supplier:  ANTIBODIES.COM