ContactUsLinkComponent
Human Apolipoprotein L1/APOL1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Apolipoprotein L1/APOL1 in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA5485-48
Supplier:  Antibodies.com
New Product
Human Apolipoprotein L1/APOL1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of human Apolipoprotein L1/APOL1 in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA5485-48
Supplier:  Antibodies.com