ContactUsLinkComponent
Human GAL12 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GAL12 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4451-96
Supplier:  ANTIBODIES.COM
New Product
Human GAL12 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GAL12 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA4451-96
Supplier:  ANTIBODIES.COM