ContactUsLinkComponent
Rat GADD45A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat GADD45A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5402-48
Supplier:  ANTIBODIES.COM
New Product
Rat GADD45A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat GADD45A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5402-48
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.