ContactUsLinkComponent
Rat Syndecan 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat Syndecan 4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4663-48
Supplier:  ANTIBODIES.COM
New Product
Rat Syndecan 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat Syndecan 4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4663-48
Supplier:  ANTIBODIES.COM