ContactUsLinkComponent
Human Glucose 6 Phosphate Dehydrogenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Glucose 6 Phosphate Dehydrogenase in serum, plasma, tissue homogenates or other e 1 * 48 Tests
Catalog # ANTIA3346-48
Supplier:  ANTIBODIES.COM
New Product
Human Glucose 6 Phosphate Dehydrogenase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Glucose 6 Phosphate Dehydrogenase in serum, plasma, tissue homogenates or other e 1 * 48 Tests
Catalog # ANTIA3346-48
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.