ContactUsLinkComponent
Human Tenomodulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Tenomodulin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5759-48
New Product
Human Tenomodulin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Tenomodulin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5759-48
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.