ContactUsLinkComponent
Human OGDH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human OGDH in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5037-48
Supplier:  ANTIBODIES.COM
New Product
Human OGDH ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human OGDH in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5037-48
Supplier:  ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.