Human IL-19 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-19 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5045-96
Supplier: ANTIBODIES.COM