ContactUsLinkComponent
Human Monkeypox Virus IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determina tion of human Monkeypox Virus IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327008-96
Supplier:  ANTIBODIES.COM
New Product
Human Monkeypox Virus IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determina tion of human Monkeypox Virus IgG in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327008-96
Supplier:  ANTIBODIES.COM