ContactUsLinkComponent
Mouse GGT1/GGT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse GGT1/GGT in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3888-96
Supplier:  ANTIBODIES.COM
New Product
Mouse GGT1/GGT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of mouse GGT1/GGT in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3888-96
Supplier:  ANTIBODIES.COM