ContactUsLinkComponent
Porcine Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of porcine Osteoprotegerin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3891-96
Supplier:  ANTIBODIES.COM
New Product
Porcine Osteoprotegerin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of porcine Osteoprotegerin in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3891-96
Supplier:  ANTIBODIES.COM