Bovine Creatine Kinase MM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of bovine Creatine Kinase MM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 96 Tests
Catalog # ANTIA3955-96
Supplier: ANTIBODIES.COM