ContactUsLinkComponent
Rat Galectin 8/Gal-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Galectin 8/Gal-8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA4447-48
Supplier:  Antibodies.com
New Product
Rat Galectin 8/Gal-8 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Galectin 8/Gal-8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA4447-48
Supplier:  Antibodies.com