Beta Lactam + Sulfanilamide + Fluoroquinolones + Tetracycline Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Beta Lactam + Sulfanilamide + Fluoroquinolones + Tetracycline in raw commingled cow&# 1 * 96 Tests
Catalog # ANTIA327183-96
Supplier: ANTIBODIES.COM