ContactUsLinkComponent
Human ZO1 Tight Junction Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human ZO1 Tight Junction Protein in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA4785-48
Supplier:  Antibodies.com
New Product
Human ZO1 Tight Junction Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of human ZO1 Tight Junction Protein in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA4785-48
Supplier:  Antibodies.com