Mouse HSP90 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse HSP90 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA5658-48
Supplier: ANTIBODIES.COM