ContactUsLinkComponent
Human TREM2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TREM2 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5065-48
New Product
Human TREM2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TREM2 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5065-48