ContactUsLinkComponent
Rat Liver Arginase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Liver Arginase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA5877-48
Supplier:  Antibodies.com
New Product
Rat Liver Arginase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of rat Liver Arginase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other bio 1 * 48 Tests
Catalog # ANTIA5877-48
Supplier:  Antibodies.com