ContactUsLinkComponent
Canine IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of canine IL-1 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA4270-48
Supplier:  Antibodies.com
New Product
Canine IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of canine IL-1 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
Catalog # ANTIA4270-48
Supplier:  Antibodies.com