ContactUsLinkComponent
Human beta Defensin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human beta Defensin 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA3743-48
Supplier:  Antibodies.com
New Product
Human beta Defensin 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human beta Defensin 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 48 Tests
Catalog # ANTIA3743-48
Supplier:  Antibodies.com