ContactUsLinkComponent
Human SARS-CoV-2 Spike Protein Total Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro qu antitative determination of human SARS-CoV-2 Spike Protein Total Antibody in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327006-96
Supplier:  Antibodies.com
New Product
Human SARS-CoV-2 Spike Protein Total Antibody ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay designed for the in vitro qu antitative determination of human SARS-CoV-2 Spike Protein Total Antibody in serum and plasma. 1 * 96 Tests
Catalog # ANTIA327006-96
Supplier:  Antibodies.com