ContactUsLinkComponent
Free Testosterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of Free Testosterone in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA326996-96
Supplier:  ANTIBODIES.COM
New Product
Free Testosterone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of Free Testosterone in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA326996-96
Supplier:  ANTIBODIES.COM