ContactUsLinkComponent
Melamine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Melamine in milk, yogurt, milk powder, egg, meat, fish, feed and other food samples. 1 * 96 Tests
Catalog # ANTIA327226-96
Supplier:  ANTIBODIES.COM
New Product
Melamine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Melamine in milk, yogurt, milk powder, egg, meat, fish, feed and other food samples. 1 * 96 Tests
Catalog # ANTIA327226-96
Supplier:  ANTIBODIES.COM