Mouse INPP4A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse INPP4A in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5122-48
Supplier: ANTIBODIES.COM
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.