ContactUsLinkComponent
Rat S100 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat S100 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA3585-48
Supplier:  Antibodies.com
New Product
Rat S100 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat S100 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fl 1 * 48 Tests
Catalog # ANTIA3585-48
Supplier:  Antibodies.com