Mouse SNAP25 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SNAP25 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4547-48
Supplier: ANTIBODIES.COM