Rat Insulin Degrading Enzyme/IDE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of rat Insulin Degrading Enzyme/IDE in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA5964-48
Supplier: Antibodies.com