Schiff reagent is intended to be used as multi-application microscopy reagent particularly in histo-cytological application prior microscopic examination. If applicable, CellaVision RAL Diagnostics recommends using the associated CellaVision RAL Diag 1 * 1 L
Catalog # RALA320680-1000
Supplier: RAL