ContactUsLinkComponent
Mouse Cytochrome P450 2E1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cytochrome P450 2E1 in tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
  ANTIA6255-48
New Product
Mouse Cytochrome P450 2E1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cytochrome P450 2E1 in tissue homogenates, cell lysates, cell culture supernates or other biolo 1 * 48 Tests
  ANTIA6255-48