ContactUsLinkComponent
Human Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of human Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 48 Tests
Catalog # ANTIA8118-48
Supplier:  Antibodies.com
New Product
Human Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determ ination of human Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biol 1 * 48 Tests
Catalog # ANTIA8118-48
Supplier:  Antibodies.com