ContactUsLinkComponent
Mouse SIRT6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SIRT6 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7385-48
New Product
Mouse SIRT6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SIRT6 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7385-48