Mouse Lysosomal Acid Lipase/LAL ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Lysosomal Acid Lipase/LAL in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA7763-48
Supplier: Antibodies.com