ContactUsLinkComponent
Mouse Free Fatty Acid Receptor 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Free Fatty Acid Receptor 3 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7643-48
Supplier:  Antibodies.com
New Product
Mouse Free Fatty Acid Receptor 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quanti tative determination of mouse Free Fatty Acid Receptor 3 in tissue homogenates, cell lysates or other biological flui 1 * 48 Tests
Catalog # ANTIA7643-48
Supplier:  Antibodies.com