ContactUsLinkComponent
Rat Ornithine Carbamoyltransferase/OTC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ornithine Carbamoyltransferase/OTC in serum, plasma, tissue homogenates, and other b 1 * 96 Tests
Catalog # ANTIA80351-96
Supplier:  ANTIBODIES.COM
New Product
Rat Ornithine Carbamoyltransferase/OTC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ornithine Carbamoyltransferase/OTC in serum, plasma, tissue homogenates, and other b 1 * 96 Tests
Catalog # ANTIA80351-96
Supplier:  ANTIBODIES.COM