Mouse IDO-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse IDO-2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7407-48
Supplier: ANTIBODIES.COM