ContactUsLinkComponent
Human TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human TGF beta 3 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6001-48
Supplier:  Antibodies.com
New Product
Human TGF beta 3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of human TGF beta 3 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6001-48
Supplier:  Antibodies.com