ContactUsLinkComponent
Human beta Casein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human beta Casein in breast milk or other biological fluids. 1 * 96 Tests
  ANTIA6774-96
New Product
Human beta Casein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human beta Casein in breast milk or other biological fluids. 1 * 96 Tests
  ANTIA6774-96
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.