ContactUsLinkComponent
Rat Calcium Pump PMCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Calcium Pump PMCA2 ATPase in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA8073-48
Supplier:  ANTIBODIES.COM
New Product
Rat Calcium Pump PMCA2 ATPase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitat ive determination of rat Calcium Pump PMCA2 ATPase in serum, plasma, tissue homogenates, cell lysates, cell culture s 1 * 48 Tests
Catalog # ANTIA8073-48
Supplier:  ANTIBODIES.COM