ContactUsLinkComponent
Mouse Neurogenin 3/NGN-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Neurogenin 3/NGN-3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6308-96
Supplier:  ANTIBODIES.COM
New Product
Mouse Neurogenin 3/NGN-3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of mouse Neurogenin 3/NGN-3 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA6308-96
Supplier:  ANTIBODIES.COM