ContactUsLinkComponent
Rat Ferritin Light Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Ferritin Light Chain in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA6268-48
Supplier:  Antibodies.com
New Product
Rat Ferritin Light Chain ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Ferritin Light Chain in serum, plasma, tissue homogenates, cell lysates, cell culture supernates 1 * 48 Tests
Catalog # ANTIA6268-48
Supplier:  Antibodies.com